Assay Details
| Gene Symbol: Masp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008555.2 | NP_032581.2 | 13 - 14 | 1799 | 73 | |
| GenBank mRNA | BC131637.1 | - | 13 - 14 | 1799 | 40109637 | 73 |
| BC131638.1 | - | 13 - 14 | 1799 | 40109639 | 73 | |
| D16492.1 | - | 1812 | 73 | |||
More Information
Set Membership: |
|

Masp1
Genomic Map