Assay Details
| Gene Symbol: Masp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008555.2 | NP_032581.2 | 14 - 15 | 1867 | 71 | |
| GenBank mRNA | BC131637.1 | - | 14 - 15 | 1867 | 40109637 | 71 |
| BC131638.1 | - | 14 - 15 | 1867 | 40109639 | 71 | |
| D16492.1 | - | 1880 | 71 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Masp1
Genomic Map