Assay Details
| Gene Symbol: Spice1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144550.4 | NP_653133.3 | 13 - 14 | 1779 | 70 | |
| GenBank mRNA | AK048789.1 | - | 13 - 14 | 1723 | 70 | |
| AY099108.1 | - | 13 - 14 | 2059 | 70 | ||
| BC024956.1 | - | 5 - 6 | 1133 | 70 | ||
| BC052381.1 | - | 13 - 14 | 1733 | 5703283 | 70 | |
| BC056960.1 | - | 13 - 14 | 1733 | 5703283 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spice1
Genomic Map