Assay Details
| Gene Symbol: Spice1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144550.4 | NP_653133.3 | 15 - 16 | 2438 | 62 | |
| GenBank mRNA | AK048789.1 | - | 15 - 16 | 2382 | 62 | |
| AY099108.1 | - | 15 - 16 | 2718 | 62 | ||
| BC024956.1 | - | 7 - 8 | 1792 | 62 | ||
| BC052381.1 | - | 15 - 16 | 2392 | 5703283 | 62 | |
| BC056960.1 | - | 15 - 16 | 2392 | 5703283 | 62 | |
More Information
Set Membership: |
|

Spice1
Genomic Map