Assay Details
| Gene Symbol: Spice1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144550.4 | NP_653133.3 | 4 - 5 | 392 | 102 | |
| GenBank mRNA | AK048789.1 | - | 4 - 5 | 336 | 102 | |
| AK165031.1 | - | 2 - 3 | 235 | 102 | ||
| AY099108.1 | - | 4 - 5 | 672 | 102 | ||
| BC052381.1 | - | 4 - 5 | 346 | 5703283 | 102 | |
| BC056960.1 | - | 4 - 5 | 346 | 5703283 | 102 | |
| CX568108.1 | - | 4 - 5 | 392 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spice1
Genomic Map