Assay Details
| Gene Symbol: Sidt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159419.1 | NP_001152891.1 | 18 - 19 | 2165 | 73 | |
| NM_198034.3 | NP_932151.2 | 18 - 19 | 2165 | 73 | ||
| GenBank mRNA | AK034344.1 | - | 3 - 4 | 315 | 73 | |
| AK046836.1 | - | 18 - 19 | 2525 | 73 | ||
| AK171920.1 | - | 18 - 19 | 73 | |||
| AY562213.1 | - | 7 - 8 | 709 | 73 | ||
| AY562214.1 | - | 7 - 8 | 709 | 73 | ||
| BC025888.1 | - | 8 - 9 | 815 | 5039343 | 73 | |
| BC079584.1 | - | 18 - 19 | 2165 | 5704672 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sidt1
Genomic Map