Assay Details
| Gene Symbol: 1700026J12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006389.1 | - | 4 - 5 | 618 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
myosin, heavy chain 15 |
Myh15 | EG667772 | -22.4kb | Anti-sense | Chr. 16 - 49057486 - 49199104 on NCBI Build GRCm38 |
| Downstream | intraflagellar transport 57 | Ift57 |
4833420A15Rik, Esrrbl1, Hippi, MHS4R2 |
474.2kb | Anti-sense | Chr. 16 - 49699294 - 49765126 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1700026J12Rik
Genomic Map