Assay Details
| Gene Symbol: Ppl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008909.2 | NP_032935.2 | 15 - 16 | 1967 | 93 | |
| GenBank mRNA | AF126834.1 | - | 15 - 16 | 1907 | 93 | |
| BC057152.1 | - | 15 - 16 | 1896 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppl
Genomic Map