Assay Details
| Gene Symbol: Eef2kmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027446.2 | NP_081722.1 | 1 - 2 | 119 | 75 | |
| GenBank mRNA | AK017533.1 | - | 1 - 2 | 120 | 75 | |
| AK133590.1 | - | 1 - 2 | 120 | 75 | ||
| AK163493.1 | - | 1 - 2 | 123 | 75 | ||
| BC039631.1 | - | 1 - 2 | 97 | 75 | ||
| BC051078.1 | - | 1 - 2 | 104 | 6311572 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC051078.1 transcripts.
Set Membership: |
|

Eef2kmt
Genomic Map