Assay Details
| Gene Symbol: Mrpl39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017404.4 | NP_059100.3 | 6 - 7 | 783 | 114 | |
| GenBank mRNA | AF239728.1 | - | 6 - 7 | 700 | 114 | |
| AK007681.1 | - | 6 - 7 | 719 | 114 | ||
| AK031942.1 | - | 6 - 7 | 704 | 114 | ||
| BC012274.1 | - | 6 - 7 | 662 | 3981658 | 114 | |
More Information
Set Membership: |
|

Mrpl39
Genomic Map