Assay Details
| Gene Symbol: Timmdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024273.2 | NP_077235.2 | 1 - 2 | 320 | 104 | |
| GenBank mRNA | AK012774.1 | - | 1 - 2 | 294 | 104 | |
| AK015460.1 | - | 1 - 2 | 298 | 104 | ||
| AK081781.1 | - | 1 - 2 | 263 | 104 | ||
| BC002243.1 | - | 1 - 2 | 282 | 3492342 | 104 | |
| CJ056343.1 | - | 1 - 2 | 322 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Timmdc1
Genomic Map