Assay Details
| Gene Symbol: Timmdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024273.2 | NP_077235.2 | 3 - 4 | 580 | 73 | |
| GenBank mRNA | AK012774.1 | - | 3 - 4 | 554 | 73 | |
| AK081781.1 | - | 3 - 4 | 523 | 73 | ||
| BC002243.1 | - | 3 - 4 | 542 | 3492342 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Timmdc1
Genomic Map