Assay Details
| Gene Symbol: Adamts1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009621.4 | NP_033751.3 | 4 - 5 | 1784 | 91 | |
| GenBank mRNA | AK156830.1 | - | 4 - 5 | 1783 | 91 | |
| AK161409.1 | - | 4 - 5 | 886 | 91 | ||
| AK163622.1 | - | 4 - 5 | 1783 | 91 | ||
| BC003269.1 | - | 4 - 5 | 1769 | 91 | ||
| BC040382.1 | - | 4 - 5 | 1768 | 3156736 | 91 | |
| BC050834.1 | - | 4 - 5 | 1768 | 6505494 | 91 | |
| D67076.1 | - | 4 - 5 | 1768 | 91 | ||
More Information
Set Membership: |
|

Adamts1
Genomic Map