Assay Details
| Gene Symbol: Fam131a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133778.2 | NP_598539.1 | 1 - 2 | 123 | 87 | |
| GenBank mRNA | AK049983.1 | - | 1 - 2 | 124 | 87 | |
| BC092532.1 | - | 1 - 2 | 154 | 6594874 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam131a
Genomic Map