Assay Details
| Gene Symbol: Ephb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010143.1 | NP_034273.1 | 13 - 14 | 2897 | 67 | |
| GenBank mRNA | AK144414.1 | - | 13 - 14 | 2914 | 67 | |
| AK158043.1 | - | 14 - 15 | 3836 | 67 | ||
| BC014822.1 | - | 13 - 14 | 2898 | 3673003 | 67 | |
| BC027805.1 | - | 9 - 10 | 2184 | 67 | ||
| BC053085.1 | - | 13 - 14 | 2897 | 5708827 | 67 | |
| X76012.1 | - | 2 - 3 | 368 | 67 | ||
| Z49086.1 | - | 13 - 14 | 2755 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephb3
Genomic Map