Assay Details
| Gene Symbol: Ephb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010143.1 | NP_034273.1 | 3 - 4 | 1214 | 68 | |
| GenBank mRNA | AK144414.1 | - | 3 - 4 | 1230 | 68 | |
| AK158043.1 | - | 4 - 5 | 2154 | 68 | ||
| BC014822.1 | - | 3 - 4 | 1215 | 3673003 | 68 | |
| BC053085.1 | - | 3 - 4 | 1214 | 5708827 | 68 | |
| U11493.1 | - | 1 - 2 | 674 | 68 | ||
| Z49086.1 | - | 3 - 4 | 1072 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephb3
Genomic Map