Assay Details
| Gene Symbol: Ephb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010143.1 | NP_034273.1 | 6 - 7 | 1837 | 88 | |
| GenBank mRNA | AK144414.1 | - | 6 - 7 | 1853 | 88 | |
| AK158043.1 | - | 7 - 8 | 2777 | 88 | ||
| BC014822.1 | - | 6 - 7 | 1838 | 3673003 | 88 | |
| BC027805.1 | - | 3 - 4 | 1020 | 88 | ||
| BC053085.1 | - | 6 - 7 | 1837 | 5708827 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ephb3
Genomic Map