Assay Details
| Gene Symbol: Tm4sf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160402.1 | NP_001153874.1 | 1 - 2 | 423 | 89 | |
| GenBank mRNA | BC147727.1 | - | 1 - 2 | 423 | 9056676 | 89 |
| BC147729.1 | - | 1 - 2 | 423 | 9056678 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC147727.1, BC147729.1, NM_001160402.1 transcripts.
Set Membership: |
|

Tm4sf19
Genomic Map