| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600007265.mm.1 |
NM_001113208.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007265.mm.1 |
NM_001113208.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007265.mm.1 |
NM_010954.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007265.mm.1 |
NM_010954.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007265.mm.1 |
TC1600000918.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600007268.mm.1 |
NM_001113208.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007268.mm.1 |
NM_001113208.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007268.mm.1 |
NM_010954.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007268.mm.1 |
NM_010954.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007268.mm.1 |
TC1600000918.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600007266.mm.1 |
NM_001113208.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007266.mm.1 |
NM_001113208.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007266.mm.1 |
NM_010954.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007266.mm.1 |
NM_010954.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007266.mm.1 |
TC1600000918.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600007269.mm.1 |
NM_001113208.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007269.mm.1 |
NM_001113208.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007269.mm.1 |
NM_010954.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007269.mm.1 |
NM_010954.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007269.mm.1 |
TC1600000918.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600007267.mm.1 |
NM_001113208.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007267.mm.1 |
NM_001113208.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007267.mm.1 |
NM_010954.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007267.mm.1 |
NM_010954.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007267.mm.1 |
TC1600000918.mm.1 |
MTA TC |