Assay Details
| Gene Symbol: Cd86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019388.3 | NP_062261.3 | 1 - 2 | 135 | 75 | |
| GenBank mRNA | AF065897.1 | - | 1 - 2 | 51 | 75 | |
| AF065898.1 | - | 1 - 2 | 51 | 75 | ||
| AF065899.1 | - | 1 - 2 | 51 | 75 | ||
| AF065900.1 | - | 1 - 2 | 51 | 75 | ||
| AK079513.1 | - | 1 - 2 | 136 | 75 | ||
| AK170054.1 | - | 1 - 2 | 136 | 75 | ||
| AK172548.1 | - | 1 - 2 | 138 | 75 | ||
| L25606.1 | - | 1 - 2 | 117 | 75 | ||
| S70108.1 | - | 1 - 2 | 100 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF065897.1, AF065898.1, AF065899.1, AF065900.1, AK079513.1, AK170054.1, AK172548.1, L25606.1, NM_019388.3, S70108.1 transcripts.
Set Membership: |
|

Cd86
Genomic Map