Assay Details
| Gene Symbol: Cd86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019388.3 | NP_062261.3 | 4 - 5 | 811 | 111 | |
| GenBank mRNA | AF065897.1 | - | 4 - 5 | 727 | 111 | |
| AF065898.1 | - | 4 - 5 | 727 | 111 | ||
| AF065899.1 | - | 4 - 5 | 727 | 111 | ||
| AF065900.1 | - | 4 - 5 | 727 | 111 | ||
| AK079513.1 | - | 4 - 5 | 812 | 111 | ||
| AK170054.1 | - | 4 - 5 | 812 | 111 | ||
| AK172548.1 | - | 4 - 5 | 814 | 111 | ||
| BC013807.1 | - | 4 - 5 | 785 | 4008635 | 111 | |
| L25606.1 | - | 4 - 5 | 793 | 111 | ||
| S70108.1 | - | 4 - 5 | 776 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd86
Genomic Map