Assay Details
| Gene Symbol: Dtx3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013371.2 | NP_001013389.2 | 3 - 4 | 2103 | 82 | |
| GenBank mRNA | AK146802.1 | - | 3 - 4 | 2011 | 82 | |
| BC023741.1 | - | 1 - 2 | 905 | 82 | ||
| BC085093.1 | - | 3 - 4 | 2008 | 30735413 | 82 | |
| BC120494.1 | - | 3 - 4 | 2044 | 82 | ||
| BC137694.1 | - | 3 - 4 | 2044 | 8860713 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dtx3l
Genomic Map