Assay Details
| Gene Symbol: Gmip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198101.1 | NP_932769.1 | 18 - 19 | 2196 | 95 | |
| GenBank mRNA | AK019874.2 | - | 7 - 8 | 946 | 95 | |
| AK052221.1 | - | 18 - 19 | 2162 | 95 | ||
| AK082337.1 | - | 18 - 19 | 2193 | 95 | ||
| AK137177.1 | - | 16 - 17 | 2189 | 95 | ||
| AK172438.1 | - | 18 - 19 | 2184 | 95 | ||
| BC057037.1 | - | 18 - 19 | 2196 | 5702855 | 95 | |
| BC060628.1 | - | 18 - 19 | 2171 | 6847400 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmip
Genomic Map