Assay Details
| Gene Symbol: Gmip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198101.1 | NP_932769.1 | 1 - 2 | 133 | 84 | |
| GenBank mRNA | AK052221.1 | - | 1 - 2 | 102 | 84 | |
| AK082337.1 | - | 1 - 2 | 130 | 84 | ||
| AK172438.1 | - | 1 - 2 | 123 | 84 | ||
| BC057037.1 | - | 1 - 2 | 133 | 5702855 | 84 | |
| BC060628.1 | - | 1 - 2 | 108 | 6847400 | 84 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC057037.1,BC060628.1,NM_198101.1 transcripts.
Set Membership: |
|

Gmip
Genomic Map