Assay Details
| Gene Symbol: Adgrg7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172825.3 | NP_766413.2 | 10 - 11 | 1446 | 94 | |
| GenBank mRNA | AK033593.1 | - | 10 - 11 | 1447 | 94 | |
| BC132302.1 | - | 10 - 11 | 1271 | 40130579 | 94 | |
| BC137963.1 | - | 10 - 11 | 1271 | 8860982 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adgrg7
Genomic Map