Assay Details
| Gene Symbol: Ydjc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076869.1 | - | 494 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK076869.1 transcripts.
Set Membership: |
|

Ydjc
Genomic Map