Assay Details
| Gene Symbol: Ydjc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026940.4 | NP_081216.1 | 1 - 2 | 186 | 74 | |
| GenBank mRNA | AK007503.1 | - | 1 - 2 | 175 | 74 | |
| AK032396.1 | - | 216 | 74 | |||
| AK076869.1 | - | 797 | 74 | |||
| BC030425.1 | - | 1 - 2 | 188 | 74 | ||
| BC115584.1 | - | 1 - 2 | 174 | 40102346 | 74 | |
| BC115585.1 | - | 1 - 2 | 174 | 40102347 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ydjc
Genomic Map