Assay Details
| Gene Symbol: Txnrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013711.3 | NP_038739.2 | 16 - 17 | 1480 | 75 | |
| GenBank mRNA | AB027566.1 | - | 16 - 17 | 1480 | 75 | |
| AF136399.1 | - | 16 - 17 | 1483 | 75 | ||
| AF171053.1 | - | 16 - 17 | 1446 | 75 | ||
| AF412308.1 | - | 15 - 16 | 1389 | 75 | ||
| AK075828.1 | - | 16 - 17 | 1458 | 75 | ||
| BC029344.1 | - | 16 - 17 | 1460 | 75 | ||
| BC052157.1 | - | 16 - 17 | 1459 | 6506286 | 75 | |
| BC052758.1 | - | 16 - 17 | 1459 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Txnrd2
Genomic Map