Assay Details
| Gene Symbol: Txnrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013711.3 | NP_038739.2 | 2 - 3 | 213 | 59 | |
| GenBank mRNA | AB027566.1 | - | 2 - 3 | 213 | 59 | |
| AF136399.1 | - | 2 - 3 | 216 | 59 | ||
| AF171053.1 | - | 2 - 3 | 179 | 59 | ||
| AF412308.1 | - | 2 - 3 | 215 | 59 | ||
| AK008004.1 | - | 2 - 3 | 132 | 59 | ||
| AK075828.1 | - | 2 - 3 | 191 | 59 | ||
| BC013688.1 | - | 2 - 3 | 193 | 3584154 | 59 | |
| BC029344.1 | - | 2 - 3 | 193 | 59 | ||
| BC052157.1 | - | 2 - 3 | 192 | 6506286 | 59 | |
| BC052758.1 | - | 2 - 3 | 192 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Txnrd2
Genomic Map