| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600010700.mm.1 |
NM_011172.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010700.mm.1 |
NM_011172.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010700.mm.1 |
TC1600001304.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600010704.mm.1 |
NM_011172.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010704.mm.1 |
NM_011172.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010704.mm.1 |
TC1600001304.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600010703.mm.1 |
NM_011172.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010703.mm.1 |
NM_011172.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010703.mm.1 |
TC1600001304.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600010702.mm.1 |
NM_011172.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010702.mm.1 |
NM_011172.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010702.mm.1 |
TC1600001304.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600010701.mm.1 |
NM_011172.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010701.mm.1 |
NM_011172.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010701.mm.1 |
TC1600001304.mm.1 |
MTA TC |