Assay Details
| Gene Symbol: Uso1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019490.1 | NP_062363.1 | 4 - 5 | 447 | 116 | |
| GenBank mRNA | AF096868.1 | - | 3 - 4 | 235 | 116 | |
| AK144638.1 | - | 4 - 5 | 438 | 116 | ||
| AK156070.1 | - | 4 - 5 | 440 | 116 | ||
| AK166587.1 | - | 5 - 6 | 407 | 116 | ||
| AK172425.1 | - | 4 - 5 | 438 | 116 | ||
| BC016069.1 | - | 4 - 5 | 447 | 4480992 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uso1
Genomic Map