Assay Details
| Gene Symbol: Cxcl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019494.1 | NP_062367.1 | 2 - 3 | 247 | 99 | |
| NR_038116.1 | 2 - 3 | 265 | 99 | |||
| GenBank mRNA | AF136449.1 | - | 2 - 3 | 247 | 99 | |
| AF178672.1 | - | 2 - 3 | 247 | 99 | ||
| AF179872.1 | - | 2 - 3 | 247 | 99 | ||
| AK020750.1 | - | 2 - 3 | 266 | 99 | ||
| AK040051.1 | - | 2 - 3 | 207 | 99 | ||
| AK050012.1 | - | 2454 | 99 | |||
| BC025903.1 | - | 2 - 3 | 235 | 5035983 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cxcl11
Genomic Map