Assay Details
| Gene Symbol: Stra6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004855.1 | - | 7 - 8 | 726 | 85 | |
| AK045033.1 | - | 8 - 9 | 1154 | 85 | ||
| AK050152.1 | - | 7 - 8 | 737 | 85 | ||
| AK050348.1 | - | 7 - 8 | 733 | 85 | ||
| BC111889.1 | - | 7 - 8 | 663 | 40060643 | 85 | |
| BC112408.1 | - | 7 - 8 | 663 | 40060646 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stra6l
Genomic Map