Assay Details
| Gene Symbol: Slc18a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153054.2 | NP_694694.1 | 10 - 11 | 103 | ||
| GenBank mRNA | AK076433.1 | - | 9 - 10 | 103 | ||
| AY779336.1 | - | 8 - 9 | 1033 | 103 | ||
| BC027192.1 | - | 10 - 11 | 1313 | 4239930 | 103 | |
| BC031146.1 | - | 10 - 11 | 1343 | 4974776 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc18a1
Genomic Map