Assay Details
| Gene Symbol: Slc18a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153054.2 | NP_694694.1 | 15 - 16 | 1788 | 149 | |
| GenBank mRNA | AK076433.1 | - | 14 - 15 | 1513 | 149 | |
| AY779336.1 | - | 13 - 14 | 1446 | 149 | ||
| BC027192.1 | - | 15 - 16 | 1726 | 4239930 | 149 | |
| BC031146.1 | - | 15 - 16 | 1756 | 4974776 | 149 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc18a1
Genomic Map