Assay Details
| Gene Symbol: Slc18a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153054.2 | NP_694694.1 | 5 - 6 | 996 | 80 | |
| GenBank mRNA | AK076433.1 | - | 4 - 5 | 720 | 80 | |
| AK144034.1 | - | 4 - 5 | 720 | 80 | ||
| AY779336.1 | - | 3 - 4 | 654 | 80 | ||
| BC027192.1 | - | 5 - 6 | 934 | 4239930 | 80 | |
| BC031146.1 | - | 5 - 6 | 964 | 4974776 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc18a1
Genomic Map