Assay Details
| Gene Symbol: Atp6v1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007509.3 | NP_031535.2 | 8 - 9 | 856 | 77 | |
| GenBank mRNA | AK146499.1 | - | 8 - 9 | 821 | 77 | |
| AK151200.1 | - | 8 - 9 | 857 | 77 | ||
| AK151322.1 | - | 8 - 9 | 829 | 77 | ||
| AK151586.1 | - | 8 - 9 | 829 | 77 | ||
| AK152718.1 | - | 8 - 9 | 829 | 77 | ||
| AK152766.1 | - | 8 - 9 | 829 | 77 | ||
| AK153341.1 | - | 8 - 9 | 820 | 77 | ||
| AK159133.1 | - | 8 - 9 | 827 | 77 | ||
| AK159153.1 | - | 8 - 9 | 827 | 77 | ||
| AK159586.1 | - | 8 - 9 | 824 | 77 | ||
| AK159701.1 | - | 8 - 9 | 828 | 77 | ||
| AK160854.1 | - | 8 - 9 | 822 | 77 | ||
| AK166669.1 | - | 8 - 9 | 796 | 77 | ||
| AK168852.1 | - | 8 - 9 | 820 | 77 | ||
| AK169155.1 | - | 8 - 9 | 835 | 77 | ||
| AK169270.1 | - | 8 - 9 | 832 | 77 | ||
| AK169420.1 | - | 8 - 9 | 824 | 77 | ||
| AK170110.1 | - | 8 - 9 | 851 | 77 | ||
| AK210829.1 | - | 3 - 4 | 275 | 77 | ||
| BC012497.1 | - | 8 - 9 | 821 | 4500843 | 77 | |
| BC046302.1 | - | 8 - 9 | 811 | 6492612 | 77 | |
| BC085300.1 | - | 8 - 9 | 803 | 6305977 | 77 | |
| Y12634.1 | - | 8 - 9 | 813 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1b2
Genomic Map