Assay Details
| Gene Symbol: Atp6v1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007509.3 | NP_031535.2 | 9 - 10 | 988 | 73 | |
| GenBank mRNA | AK146499.1 | - | 9 - 10 | 953 | 73 | |
| AK151200.1 | - | 9 - 10 | 989 | 73 | ||
| AK151322.1 | - | 9 - 10 | 961 | 73 | ||
| AK151586.1 | - | 9 - 10 | 961 | 73 | ||
| AK152718.1 | - | 9 - 10 | 961 | 73 | ||
| AK152766.1 | - | 9 - 10 | 961 | 73 | ||
| AK153341.1 | - | 9 - 10 | 952 | 73 | ||
| AK159133.1 | - | 9 - 10 | 959 | 73 | ||
| AK159153.1 | - | 9 - 10 | 959 | 73 | ||
| AK159586.1 | - | 9 - 10 | 956 | 73 | ||
| AK159701.1 | - | 9 - 10 | 960 | 73 | ||
| AK160854.1 | - | 9 - 10 | 954 | 73 | ||
| AK166669.1 | - | 9 - 10 | 928 | 73 | ||
| AK168852.1 | - | 9 - 10 | 952 | 73 | ||
| AK169155.1 | - | 9 - 10 | 967 | 73 | ||
| AK169270.1 | - | 9 - 10 | 964 | 73 | ||
| AK169420.1 | - | 9 - 10 | 956 | 73 | ||
| AK170110.1 | - | 9 - 10 | 983 | 73 | ||
| AK198089.1 | - | 1 - 2 | 56 | 73 | ||
| AK201716.1 | - | 1 - 2 | 56 | 73 | ||
| AK203319.1 | - | 1 - 2 | 56 | 73 | ||
| AK204547.1 | - | 1 - 2 | 56 | 73 | ||
| BC012497.1 | - | 9 - 10 | 953 | 4500843 | 73 | |
| BC046302.1 | - | 9 - 10 | 943 | 6492612 | 73 | |
| BC085300.1 | - | 9 - 10 | 935 | 6305977 | 73 | |
| Y12634.1 | - | 9 - 10 | 73 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1b2
Genomic Map