Assay Details
| Gene Symbol: Pknox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016670.3 | NP_057879.2 | 4 - 5 | 515 | 70 | |
| NR_027493.1 | 5 - 6 | 762 | 70 | |||
| GenBank mRNA | AK131788.1 | - | 3 - 4 | 544 | 70 | |
| AK134000.1 | - | 5 - 6 | 672 | 70 | ||
| BC052701.1 | - | 3 - 4 | 410 | 5721441 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pknox1
Genomic Map