Assay Details
| Gene Symbol: Pknox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016670.3 | NP_057879.2 | 8 - 9 | 1019 | 98 | |
| NR_027493.1 | 9 - 10 | 1266 | 98 | |||
| GenBank mRNA | AF061270.1 | - | 8 - 9 | 991 | 98 | |
| AK131788.1 | - | 7 - 8 | 1048 | 98 | ||
| AK134000.1 | - | 9 - 10 | 1176 | 98 | ||
| BC052701.1 | - | 7 - 8 | 914 | 5721441 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pknox1
Genomic Map