Assay Details
| Gene Symbol: Pknox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016670.3 | NP_057879.2 | 9 - 10 | 1094 | 111 | |
| NR_027493.1 | 10 - 11 | 1341 | 111 | |||
| GenBank mRNA | AF061270.1 | - | 9 - 10 | 1066 | 111 | |
| AK131788.1 | - | 8 - 9 | 1123 | 111 | ||
| AK134000.1 | - | 10 - 11 | 1251 | 111 | ||
| BC052701.1 | - | 8 - 9 | 989 | 5721441 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pknox1
Genomic Map