Assay Details
| Gene Symbol: Fam214b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253353.1 | NP_001240282.1 | 7 - 8 | 1845 | 79 | |
| NM_001253354.1 | NP_001240283.1 | 8 - 9 | 1622 | 79 | ||
| NM_172691.3 | NP_766279.1 | 7 - 8 | 1626 | 79 | ||
| NR_045567.1 | 6 - 7 | 1567 | 79 | |||
| GenBank mRNA | AK045811.1 | - | 7 - 8 | 1610 | 79 | |
| AK149610.1 | - | 7 - 8 | 1703 | 79 | ||
| AK154748.1 | - | 6 - 7 | 1961 | 79 | ||
| AK161380.1 | - | 7 - 8 | 1553 | 79 | ||
| AK173197.1 | - | 4 - 5 | 371 | 79 | ||
| BC044832.1 | - | 6 - 7 | 1783 | 79 | ||
| BC046771.1 | - | 8 - 9 | 1612 | 5703628 | 79 | |
| BC054736.1 | - | 6 - 7 | 1567 | 79 | ||
| BC071191.1 | - | 5 - 6 | 1314 | 6467750 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam214b
Genomic Map