Assay Details
| Gene Symbol: Spata31d1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177711.3 | NP_808379.2 | 1 - 2 | 237 | 56 | |
| GenBank mRNA | AK077018.1 | - | 1 - 2 | 237 | 56 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077018.1 transcripts.
Set Membership: |
|

Spata31d1d
Genomic Map