Assay Details
| Gene Symbol: Spata31d1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177711.3 | NP_808379.2 | 3 - 4 | 333 | 75 | |
| GenBank mRNA | AK077018.1 | - | 3 - 4 | 333 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077018.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533265 | AK077018.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533265 | NM_177711.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533265 | AK077018.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533265 | NM_177711.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623726 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 281196 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782025 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 143734 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138384 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533265 | AK077018.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533265 | NM_177711.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533265 | AK077018.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533265 | NM_177711.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081227 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07893675 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138384 | - | - |
| Whole Mouse Genome G4122A | A_52_P533265 | AK077018.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533265 | NM_177711.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533265 | AK077018.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533265 | NM_177711.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123556 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114423 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10565037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10827599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10592575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10561258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10565036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10638622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10561280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10827598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10565030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10565004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10638620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10827595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10565035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10561264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081139 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10827566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10682099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10827568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10827590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081251 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10827563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10682064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10872399 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081278 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10409791 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409795 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409794 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409792 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409793 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438584_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10409790 | - | - |
| Mouse Genome 430 2.0 Array | 1438584_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438584_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 238663_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 238663_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300013615.mm.1 | NM_177711.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013615.mm.1 | NM_177711.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013615.mm.1 | TC1300002123.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438584_at | - | - |
| Mouse Expression Set 430 | 1438584_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438584_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438584_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 366674 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5860537 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5860537 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005860537 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2776773 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2776773 | - | - |
| mouse-6 v1.1 (Illumina) | scl43876.4.1_55-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5860537 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl43876.4.1_55-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5860537 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2997695 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2997695 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2776775 | - | - |

Spata31d1d
Genomic Map