| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100025791.mm.1 |
NM_001159410.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025791.mm.1 |
NM_001159410.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025791.mm.1 |
NM_008193.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025791.mm.1 |
NM_008193.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025791.mm.1 |
TC1100002844.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100025789.mm.1 |
NM_001159410.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025789.mm.1 |
NM_001159410.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025789.mm.1 |
NM_008193.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025789.mm.1 |
NM_008193.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025789.mm.1 |
TC1100002844.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100025794.mm.1 |
NM_001159410.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025794.mm.1 |
NM_001159410.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025794.mm.1 |
TC1100002844.mm.1 |
MTA TC |