Assay Details
| Gene Symbol: Guk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159410.1 | NP_001152882.1 | 4 - 5 | 294 | 91 | |
| NM_008193.3 | NP_032219.2 | 4 - 5 | 457 | 91 | ||
| GenBank mRNA | AK004205.1 | - | 4 - 5 | 316 | 91 | |
| AK013514.1 | - | 4 - 5 | 320 | 91 | ||
| BC024625.1 | - | 4 - 5 | 321 | 4485215 | 91 | |
| CO429095.1 | - | 4 - 5 | 283 | 91 | ||
| EL957688.1 | - | 4 - 5 | 301 | 91 | ||
| U53514.1 | - | 4 - 5 | 277 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Guk1
Genomic Map