Assay Details
| Gene Symbol: Sept11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009818.1 | NP_001009818.1 | 1 - 2 | 136 | 88 | |
| GenBank mRNA | AK028475.1 | - | 1 - 2 | 68 | 88 | |
| AK166517.1 | - | 1 - 2 | 157 | 88 | ||
| AK171368.1 | - | 1 - 2 | 157 | 88 | ||
| BC031456.1 | - | 1 - 2 | 43 | 4037515 | 88 | |
| BC062206.1 | - | 1 - 2 | 177 | 5150858 | 88 | |
| BC064466.1 | - | 1 - 2 | 136 | 6825549 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028475.1, AK166517.1, AK171368.1, BC031456.1, BC062206.1, BC064466.1, NM_001009818.1 transcripts.
Set Membership: |
|

Sept11
Genomic Map