Assay Details
| Gene Symbol: Sept11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009818.1 | NP_001009818.1 | 4 - 5 | 641 | 64 | |
| GenBank mRNA | AK136063.1 | - | 2 - 3 | 270 | 64 | |
| AK166517.1 | - | 4 - 5 | 662 | 64 | ||
| AK170945.1 | - | 3 - 4 | 451 | 64 | ||
| AK171368.1 | - | 4 - 5 | 662 | 64 | ||
| BC031456.1 | - | 4 - 5 | 548 | 4037515 | 64 | |
| BC062206.1 | - | 4 - 5 | 682 | 5150858 | 64 | |
| BC064466.1 | - | 4 - 5 | 641 | 6825549 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sept11
Genomic Map