Assay Details
| Gene Symbol: Sept11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009818.1 | NP_001009818.1 | 6 - 7 | 892 | 72 | |
| GenBank mRNA | AK028475.1 | - | 6 - 7 | 824 | 72 | |
| AK166517.1 | - | 6 - 7 | 913 | 72 | ||
| AK170945.1 | - | 5 - 6 | 702 | 72 | ||
| AK171368.1 | - | 6 - 7 | 913 | 72 | ||
| BC031456.1 | - | 6 - 7 | 799 | 4037515 | 72 | |
| BC062206.1 | - | 6 - 7 | 933 | 5150858 | 72 | |
| BC064466.1 | - | 6 - 7 | 892 | 6825549 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sept11
Genomic Map