Assay Details
| Gene Symbol: Art3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181728.2 | NP_859417.2 | 7 - 8 | 1096 | 68 | |
| GenBank mRNA | AJ311769.1 | - | 5 - 6 | 909 | 68 | |
| AJ311771.1 | - | 5 - 6 | 909 | 68 | ||
| AJ311773.1 | - | 5 - 6 | 909 | 68 | ||
| AJ311774.1 | - | 4 - 5 | 200 | 68 | ||
| BC141069.1 | - | 5 - 6 | 956 | 9055681 | 68 | |
| BC145327.1 | - | 5 - 6 | 956 | 9053883 | 68 | |
| BE625316.1 | - | 4 - 5 | 221 | 68 | ||
More Information
Set Membership: |
|

Art3
Genomic Map